Learn More
Masterflex™ Tubo per pompa in silicone arrotolato Masterflex™
Codice prodotto. 11715158
Click to view available options
Dimensioni dei tubi:
13
14
15
16
18
24
25
35
37
Dimensione della confezione:
400ft
Pezzo
Codice prodotto. 11715158
Marca: Masterflex™ 9640313
Attualmente non disponibile: Non abbiamo informazioni su quando questo prodotto sarà disponibile.
Visualizzare prodotti alternativi
Reduce scrap with spooled tubing. Masterflex™ L/S™ Platinum-Cured Silicone Pump Tubing features a smooth surface and low protein binding levels, ideal for pharmaceutical, biotechnology, peristaltic pumping and food applications.
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
  • Ensure that you always have enough tubing on hand
  • Slightly greater clarity
  • Preferred by pharmaceutical and biotechnology industries
  • Fewer potential leachables
  • USP Class VI
  • FDA compliant
Specifica
Certificazione/Conformità FDA approved; Prop 65 NA (no Prop 65 chemicals)
Diametro (sistema metrico) interno 0.8 mm
Materiale Platinum-cured Silicone
Overall Length (English) 500 ft.
Overall Length (Metric) 152.4 m
Min. Operating Temperature (Metric) °
Dimensioni dei tubi 13
Portata 36 mL/min.
Flessibile flangiato 1/16 in.
Metodo di sterilizzazione Gamma Irradiation
Pressione massima 40 psi, 2.7 bar
Livello di vuoto 660 mm Hg
Max. Operating Temperature (Metric) 100°C
Product Type Pump Tubing