Learn More
Masterflex™ Tubo per pompa in silicone arrotolato Masterflex™
Click to view available options
Dimensioni dei tubi:
13
14
15
16
18
24
25
35
37
missing translation for 'unitSize'
400ft
Pezzo
Description
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
- Ensure that you always have enough tubing on hand
- Slightly greater clarity
- Preferred by pharmaceutical and biotechnology industries
- Fewer potential leachables
- USP Class VI
- FDA compliant
Spécification
Spécification
| Diametro (sistema metrico) interno | 1.6 mm |
| Materiale | Platinum-cured Silicone |
| Overall Length (Metric) | 15,200 cm |
| Da utilizzare con (applicazione) | Pharmaceutical and biotechnology industries |
| Min. Operating Temperature (Metric) | ° |
| Quantità confezione | 1 Ea. |
| Dimensioni dei tubi | 14 |
| Portata | 0.21 to 130 mL/min. |
| Flessibile flangiato | 1/16 in. |
| Pressione massima | 40 psi |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu