Learn More
ME2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24944-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ME2 encodes a mitochondrial NAD-dependent malic enzyme, a homotetrameric protein, that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.Specifica
| ME2 | |
| Polyclonal | |
| Unconjugated | |
| ME2 | |
| Malic enzyme 2, ME2, NAD ME | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4200 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P23368 | |
| ME2 | |
| ME2 Fusion Protein Ag20741 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |