missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15196-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MED10 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.Specifica
| MED10 | |
| Polyclonal | |
| Unconjugated | |
| MED10 | |
| L6, MED10, mediator complex subunit 10, NUT2, TRG 17, TRG 20, TRG17, TRG20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 28077, 290939, 84246 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BTT4, Q9CXU0 | |
| Med10 | |
| MED10 Fusion Protein Ag7336 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto