missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15196-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MED10 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.Specifica
| MED10 | |
| Polyclonal | |
| Unconjugated | |
| MED10 | |
| L6, MED10, mediator complex subunit 10, NUT2, TRG 17, TRG 20, TRG17, TRG20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 28077, 290939, 84246 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BTT4, Q9CXU0 | |
| Med10 | |
| MED10 Fusion Protein Ag7336 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion