missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5109952
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: PYFEGLRKHY LLPAIEYFNG GPPAETDFGG DYGGTQYSLV VFNTVDCAPE SYVQCHAPTS SAYEFVTWLD GIKFMGGGGE SCSLIAEGLS TALQLFDDFK KMREQIGQTH RVCLLICNSP PYLLPAVEST TYSGCTTENL VQQIGERGIH FSIVSPRKLP ALRLLFEKAA P.
This gene encodes a component of the transcriptional coactivator complex termed the Mediator complex. This complex is required for transcription of most RNA polymerase II-dependent genes. The encoded protein plays a role in chromatin modification and in preinitiation complex assembly. Mutations in this gene are associated with Charcot-Marie-Tooth disease type 2B2.
Specifica
| MED25 | |
| Polyclonal | |
| Unconjugated | |
| MED25 | |
| 2610034E13Rik; 2610529E18Rik; ACID1; Activator interaction domain-containing protein 1; activator-recruited cofactor 92 kDa component; ARC/mediator transcriptional coactivator subunit; ARC92; BVSYS; CMT2B2; DKFZp434K0512; ESTM2; hypothetical protein LOC406793; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 25; MED25; Mediator complex subunit 25; mediator of RNA polymerase II transcription subunit 25; mediator of RNA polymerase II transcription, subunit 25 homolog; mediator of RNA polymerase II transcription, subunit 9 homolog; MGC70671; mMED25; P78; prostate-derived protein 78 kDa; PTOV2; RGD1305137; TCBAP0758; zgc:63555 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 292889, 75613, 81857 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.64 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q71SY5, Q8VCB2 | |
| MED25 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-200 of human MED25 (NP_112235.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto