Learn More
Ansell MEDI-GRIP Guanti chirurgici in lattice
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Dimensione della confezione:
50 paia
Descrizione
- Proprietary anatomic-shaped glove formers to provide a comfortable fit and minimizes hand fatigue
- Soft, malleable latex formulation with optimized thickness to provide good tactile sensitivity and dexterity
- Polymer coated for fast and easy donning
Specifica
Specifica
| AQL | 0.65 |
| Colore | Natural |
| Monouso | Single-use |
| Finitura esterna | Fully Textured |
| Materiale | Natural Rubber Latex |
| Powdered | Powder Free |
| Size | 5.5 |
| Sterilità | Sterile |