missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Media Bottle PET With Cap
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Capacità (sistema metrico):
125 mL
250 mL
500 mL
1000 mL
Dimensione della confezione:
pezzi 24
pezzi 48
pezzi 60
pezzi 96
Descrizione
- Fisherbrand Media Bottles - These bottles are engineered to provide a comprehensive solution for storing media, biological buffers and a variety of reagents
- These bottles are designed to be leak-proof, shatter proof and freezer-safe, making them ideal for storing and transporting valuable samples
- Fisherbrand Media Bottles are the perfect choice for a reliable and convenient storage solution that meets the rigorous demands of modern laboratories
- Fisherbrand's media bottles are the perfect blend of durability, safety and versatility
Specifica
Specifica
| Capacità (sistema metrico) | 1000 mL |
| Capacità (sistema imperiale) | 0.264 gal. |
| Colore | Clear |
| Materiale | PET |
| Sterilità | Sterile |
| Closure Included | Yes |
| Monouso | Single-use |
| Graduated | Yes |
| Bocca | Threaded |
| Forma | Square |
| Vedi altri risultati |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering