Learn More
DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Rubber Cap/Liner
Sfoglia Tutto Wheaton Science Products Prodotti
Click to view available options
Capacità (sistema metrico):
125 mL
250 mL
500 mL
Quantità:
24 EA
48 EA
Dimensione della confezione:
pezzi 24
pezzi 48
Descrizione
- Manufactured from borosilicate glass which provides the best chemical resistance
- Clear borosilicate glass bottle
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
- Black Phenolic Rubber Lined Cap
- Hazardous communication label included with each case of bottles allows for content ID and safety notification
- Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Specifica
Specifica
| Capacità (sistema metrico) | 125 mL |
| Colore | Clear |
| Materiale | Borosilicate Glass (Type I) |
| Quantità | 48 EA |
| Colore chiusura | Black |
| Product Type | Media Bottle |
| Rivestimento interno chiusura | Rubber |
| Materiale chiusura | Phenolic |