Learn More
Medical Viscosity Standard, 10.0 mPa.s (25.00°C ), Paragon Scientific™
Codice prodotto. 16000230
Click to view available options
Quantità:
100 mL
500 mL
Dimensione della confezione:
100mL
500mL
Codice prodotto. 16000230
Marca: Paragon Scientific MGVS100500
Tested in strict accordance with ASTM D2162. These medical grade viscosity standards are fully traceable to national standards and are also test equipment compatible. Typically used, but not limited to, the calibration and verification of viscosity measuring equipment.
  • Fully traceable to international standards
  • Certified in accordance to ISO 17025 & ISO 17034 under Paragon's UKAS accreditation
  • Certified in strict accordance with ASTM D2162
  • Tamper evident security packaging
  • 12 month shelf life
  • Manufactured in the United Kingdom
  • Contains antibacterial agent
Specifica
Certificazione/Conformità ISO 17025, ISO 17034
Quantità 500 mL
Nominal values quoted. Certified Values will vary from batch to batch.