Learn More
Fisherbrand™ Meker Burner, Natural Gas
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Diametro (sistema metrico) ingresso esterno:
24 mm
30 mm
Altezza (sistema metrico):
162 mm
192 mm
Dimensione della confezione:
Pezzo
Descrizione
- The hottest flame temperature for all natural gas burners is between 900° and 1000°C at the tip of the blue flame cone
- Constructed utilizing a heavy pressure diecast base painted in a blue chemically resistant acrylic finish
- All burners tested for flame quality and leakage prior to packing
- Tests are carried out at the standard gas pressure for each specific type gas
- Temperatures dependent on mains pressure and the chemical construction of the gas which varies in different parts of the world
- Standard mains pressure for natural gas is 20 millibars
- Not fitted with an air regulator
Specifica
Specifica
| Product Type | Meker Burner |
| Tipo di gas | Natural Gas |
| Diametro (sistema metrico) ingresso esterno | 30 mm |
| Altezza (sistema metrico) | 192 mm |
| N. per confezione | 1 Ea. |