Learn More
Menthone (Mixture of Diastereomers), TRC
Product Code. 30010729
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010729 50 g 50g
30005381 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30010729 Supplier Toronto Research Chemicals Supplier No. TRCM21913050G
 Restricted Item
Spedizione stimata: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10458-14-7
Molecular Formula C10 H18 O
Molecular Weight (g/mol) 154.25
Synonym Menthone, Cyclohexanone, 5-methyl-2-(1-methylethyl)- (9CI, ACI), 5-Methyl-2-(1-methylethyl)cyclohexanone (ACI), p-Menthan-3-one (8CI), 2-Isopropyl-5-methylcyclohexanone, 5-Methyl-2-isopropylcyclohexanone, NSC 113134, NSC 9280
IUPAC Name 5-methyl-2-propan-2-ylcyclohexan-1-one
SMILES CC(C)C1CCC(C)CC1=O
Specifications
Quantity 50 g
InChI Formula InChI=1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3
Formula Weight 154.1358
Chemical Name or Material Menthone