missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEP1A Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA547467
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse MEP1A and recombinant human MEP1B is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
Specifica
| MEP1A | |
| Polyclonal | |
| Unconjugated | |
| MEP1A | |
| AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; endopeptidase-24.18 subunit alpha; Mep1; Mep-1; MEP1A; Mep-1a; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4224 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q16819 | |
| MEP1A | |
| Mouse myeloma cell line NS0-derived recombinant human Meprin alpha Subunit/MEP1A Val22-Gln601. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto