Description
Cadherin-8 Monoclonal specifically detects Cadherin-8 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Dilution | ELISA |
| Product Type | Antibody |
| Form | Purified |
| Product Line | Novus |