missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Metal Detectable Bottles

Codice prodotto. p-7239886 Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Capacità (sistema metrico):
250 mL
500 mL
Unit Size:
pezzi 4
This item is not returnable. View return policy

Product Code. 17534726

Brand: Sampling Systems™ M1047B250

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Metal Detectable Bottles are made from food grade and EU compliant PP. The robust design means they are ideal for powders and most liquid applications. The lid screws on giving a tight connection.

  • Our metal detectable tubs are ideal for use in manufacturing in food companies and general industry.
  • The bottles have a wide opening and so are ideal for use with powders.
  • The range is manufactured from a special grade of metal detectable polypropylene (PP).
  • Polypropylene has very good chemical resistance.
  • The whole range is manufactured in cleanroom conditions and packed in a ISO Class 7 (10,000) cleanroom.
  • This ensures the cleanliness of the product.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • Simply unwrap and use.
  • They are ideal for use with powders, granules and pastes.
  • The blue color means they can easily seen if left in the material.
  • The containers are available in 250 mL and 500 mL sizes.
  • The containers are powder tight and splash tight.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • 250 mL volume is the standard size.

Specifications

Capacità (sistema metrico) 250 mL
Colore Blue
Materiale Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.