Learn More
Sampling Systems™ Metal Detectable Spoons
Codice prodotto. 17504716 Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Capacità (sistema metrico):
10 mL
Dimensione della confezione:
pezzi 4
Codice prodotto. 17504716
Marca: Sampling Systems™ M1006B10
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Metal Detectable Spoons have a robust design and this means they can be used repeatedly however for demanding applications they may be used as a single-use spoon.
  • Our Metal Detectable Spoons are made from food grade polypropylene (PP).
  • A special food grade filler is mixed with the PP and this makes the final product both metal detectable and visible by X-rays.
  • All products in this metal detectable range are moulded and packed in a cleanroom.
  • This ensures the cleanliness of the product.
  • Simply unwrap and use.
  • They are ideal for use with powders, granules, liquids and pastes.
  • The range can be easily cleaned after use.
  • There are no dead spaces or crevices which can lead to bacteria traps.
  • Made from high quality PP.
  • Conforms to latest EU food and FDA requirements.
  • Just a small part of metal detectable range of products.
Specifica
Capacità (sistema metrico) 10 mL
Colore Blue