Learn More
MP Biomedicals™ Methionine/Choline Control Diet, Pellet
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantità:
10 kg
20 kg
50 kg
Dimensione della confezione:
10kg
20kg
50kg
Descrizione
- Methionine/choline control diet (MCCD) triggers the resolution of hepatic inflammatory and fibrotic reactions and hepatocyte apoptosis, suggesting that MCDD-induced steatohepatitis is also reversible
- CD diet is used in Non-alcoholic steatohepatitis study
Specifica
Specifica
| Product Type | Animal Diet |
| Formato | Pellets |
| Quantità | 50 kg |
| Requisiti di stoccaggio | Room Temperature |