Learn More
Methyl 4,4,4-trifluoroacetoacetate, 95%
Codice prodotto. p-7045286
Click to view available options
Quantità:
5 g
25 g
Dimensione della confezione:
25g
5g
Codice prodotto. 11348255
Marca: Thermo Scientific Alfa Aesar L10140.06
Richiesta di formato massivo o personalizzato
CAS: 83643-84-9 | C5H5F3O3 | 170.087 g/mol
It is used as a drug intermediate. It is a cyclization, oxidation and halogenating reaction catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a drug intermediate. It is a cyclization, oxidation and halogenating reaction catalyst.
Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 83643-84-9
Formula molecolare C5H5F3O3
Molecular Weight (g/mol) 170.087
Numero MDL MFCD00041004
InChI Key LKMUBWWZTSZGGV-UHFFFAOYSA-N
PubChem CID 550266
IUPAC Name methyl 4,4,4-trifluoro-3-oxobutanoate
SMILES COC(=O)CC(=O)C(F)(F)F
Specifica
Materiale o nome chimico Methyl 4,4,4-trifluoroacetoacetate
Densità 1.34
Punti di ebollizione 117°C to 118°C
Punto d'infiammabilità 40°C (104°F)
Indice di rifrazione 1.372
Quantità 5 g
Numero UN UN3272
Beilstein 1777343
Sinonimo methyl trifluoroacetoacetate, methyl 4,4,4-trifluoroacetoacetate, 4,4,4-trifluoroacetoacetic acid methyl ester, methyl-4,4,4-trifluoro-3-oxobutanoate, butanoic acid, 4,4,4-trifluoro-3-oxo-, methyl ester, acmc-209psv, methyltri-fluoroacetoacetate, methyl4,4,4-trifluoroacetoacetate, methyl-4,4,4-trifluoroacetoacetate, methyl 3-oxo-4,4,4-trifluorobutyrate
Peso formulazione 170.09
Percent Purity 95%