Learn More
Methyl 4-iodo-2-methoxybenzoate, 98+%
Product Code. 15453287
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453287 25 g 25g
15443287 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15453287 Supplier Thermo Scientific Alfa Aesar Supplier No. B25529.14
 Restricted Item
In stock
Add to basket
CAS: 148490-97-5 | C9H9IO3 | 292.072 g/mol
Intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Intermediate in organic syntheses.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from light, oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 148490-97-5
Molecular Formula C9H9IO3
Molecular Weight (g/mol) 292.072
MDL Number MFCD06797865
InChI Key MCNOTXROWOGSGU-UHFFFAOYSA-N
Synonym benzoic acid,4-iodo-2-methoxy-, methyl ester, 4-iodo-2-methoxybenzoic acid methyl ester, 4-iodo-2-methoxy-benzoic acid methyl ester, pubchem4974, acmc-20adk9, methyl4-iodo-2-methoxybenzoate, methyl-4-iodo-2-methoxybenzoate, 2-methoxy-4-iodo-benzoic acid methyl ester
PubChem CID 22619468
IUPAC Name methyl 4-iodo-2-methoxybenzoate
SMILES COC1=C(C=CC(=C1)I)C(=O)OC
Specifications
Melting Point 44°C to 46°C
Quantity 25 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 292.07
Percent Purity ≥98%
Chemical Name or Material Methyl 4-iodo-2-methoxybenzoate