Learn More
Metro™ Double-Deep qwikTRAK™ Track Sets
Codice prodotto. 15927001
Click to view available options
Profondità (sistema imperiale):
30 in.
72 in.
84 in.
96 in.
108 in.
120 in.
132 in.
144 in.
156 in.
168 in.
180 in.
192 in.
204 in.
216 in.
228 in.
240 in.
252 in.
Peso (sistema imperiale):
4 lb.
9 lb.
10 lb.
11 lb.
13 lb.
14 lb.
15 lb.
17 lb.
18 lb.
19 lb.
21 lb.
22 lb.
23 lb.
25 lb.
26 lb.
27 lb.
29 lb.
Dimensione della confezione:
Pezzo
Codice prodotto. 15927001
Marca: Metro™ LBTS17NA
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Create tracks up to 21 ft.
  • For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q and MetroMax i shelving systems
  • Sets include tracks and hardware needed to assemble system up to 21 ft. (640cm)
  • 17-823-756 (LBTS2.5NA) includes a joining kit to connect the track to other track sets
Specifica
Product Type Track Set
Profondità (sistema imperiale) 204 in.
Profondità (sistema metrico) 518.16 cm
Da utilizzare con (apparecchiatura) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q™ and MetroMax i™ shelving system
Include Tracks and hardware needed to assemble double deep qwikTRAK™ system up to 21 ft.
Peso (sistema imperiale) 23 lb.
Peso (sistema metrico) 10.5 kg