Learn More
Metro™ HD Super Stainless Lab Worktable, Stationary
Click to view available options
Dimensioni (L x P x A):
48 x 30 x 34 in.
60 x 30 x 34 in.
60 x 36 x 34 in.
70 x 30 x 34 in.
72 x 30 x 34 in.
96 x 30 x 34 in.
Materiale:
Stainless Steel
Stainless Steel (14 ga.)
Include:
Bottom 3-sided frame (SS)
Bottom H-frame (SS)
Bottom shelf (SS)
Dimensione della confezione:
Pezzo
Descrizione
- 34 in. high (86.4cm) seamless, 14-gauge stainless-steel work surface with rounded edges
- Support structure virtually eliminates drumming
- Posts with adjustable feet compensate for slight floor irregularities
- Leveling foot on each post
Order Info
Shipped knocked down. Assemble easily without tools. Other sizes available; call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifica
Specifica
| Product Type | Stationary Table |
| Stile della base | Open with side and rear crossrails |
| Certificazione/Conformità | NSF |
| Dimensioni (L x P x A) | 70 x 30 x 34 in. |
| Da utilizzare con (applicazione) | Ideal for lab |
| Altezza (sistema imperiale) | 34 in. |
| Altezza (sistema metrico) | 86.36 cm |
| Include | Bottom 3-sided frame (SS) |
| Lunghezza (sistema imperiale) | 30 in. |
| Lunghezza (sistema metrico) | 76.2 cm |
| Vedi altri risultati |