Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Product Code. 15986591
Click to view available options
Larghezza (sistema imperiale):
14 in.
18 in.
21 in.
24 in.
Lunghezza (sistema imperiale):
24 in.
30 in.
36 in.
42 in.
48 in.
60 in.
Unit Size:
Pezzo
Product Code. 15986591
Brand: Metro™ 1836NFS
Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.
  • All-stainless construction
  • Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Product Type Solid Shelf
Colore Silver
Materiale Stainless Steel
Autoclavabile Autoclavable
Dimensioni (L x P) 36 x 18 in. (91.4 x 45.7 cm)
Da utilizzare con (apparecchiatura) Metro Super Erecta Shelving System
Lunghezza (sistema imperiale) 36 in.
Lunghezza (sistema metrico) 91.4 cm
Larghezza (sistema imperiale) 18 in.
Larghezza (sistema metrico) 45.7 cm