Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Click to view available options
Dimensioni (L x P):
14 x 30 in.
14 x 36 in.
14 x 48 in.
14 x 60 in.
14 x 72 in.
18 x 24 in.
18 x 30 in.
18 x 36 in.
18 x 54 in.
18 x 60 in.
21 x 24 in.
21 x 30 in.
21 x 36 in.
21 x 42 in.
21 x 48 in.
21 x 54 in.
21 x 72 in.
24 x 24 in.
24 x 30 in.
24 x 36 in.
24 x 42 in.
24 x 48 in.
24 x 54 in.
30 x 48 in.
30 x 60 in.
30 x 72 in.
36 x 48 in.
36 x 60 in.
36 x 72 in.
Dimensione della confezione:
Pezzo
Descrizione
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Corrosion resistant finish for wet or high humidity environments
- Antimicrobial is built into finish to keep product clean
Specifica
Specifica
| Product Type | Wire Shelf |
| Dimensioni (L x P) | 14 x 48 in. |
| Finitura | Metroseal 3 with Microban |