Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Codice prodotto. p-4683754
Click to view available options
Dimensioni (L x P):
35.5 x 182.9 in. (14 x 72 cm)
36 x 60 in. (91 x 152.4 cm)
45.7 x 121.9 in. (18 x 48 cm)
45.7 x 183 in. (18 x 72 cm)
45.7 x 61 in. (18 x 24 cm)
45.7 x 91.4 in. (18 x 36 cm)
53 x 106.6 in. (21 x 42 cm)
53 x 21.9 in. (21 x 48 cm)
53 x 37 in. (21 x 54 cm)
53 x 63 in. (28 x 28 cm)
53 x 76 in. (30 x 30 cm)
61 x 106.6 in. (24 x 42 cm)
61 x 121.9 in. (24 x 48 cm)
61 x 137 in. (24 x 54 cm)
61 x 152.4 in. (24 x 60 cm)
76 x 121.9 in. (30 x 48 cm)
76 x 182.9 in. (30 x 72 cm)
76 x 91.4 in. (30 x 36 cm)
91 x 182.9 in. (36 x 72 cm)
91 x 91 in. (36 x 36 cm)
Profondità (sistema metrico):
355 mm
457 mm
530 mm
610 mm
760 mm
910 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15946611
Marca: Metro™ 1872NS
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifica
Product Type Wire Shelf
Materiale Stainless Steel
Profondità (sistema imperiale) 18 in.
Profondità (sistema metrico) 457 mm
Dimensioni (L x P) 45.7 x 183 in. (18 x 72 cm)
Da utilizzare con (apparecchiatura) Metro Super Erecta Shelving System
Include Plastic sleeves and wedges
Lunghezza (sistema imperiale) 72 in.
Lunghezza (sistema metrico) 182.9 cm
Larghezza (sistema imperiale) 18 in.
Larghezza (sistema metrico) 45.7 cm