Learn More
Metro™ Wire Shelving Track Sets
Codice prodotto. 15927311
Click to view available options
Lunghezza (sistema imperiale):
27 in.
72 in.
84 in.
96 in.
108 in.
120 in.
132 in.
144 in.
156 in.
168 in.
180 in.
192 in.
204 in.
216 in.
228 in.
240 in.
252 in.
Dimensione della confezione:
Pezzo
Codice prodotto. 15927311
Marca: Metro™ TTS14NA
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For use with Metro Super Erecta™, Super Erecta Pro™ and Super Adjustable™ Top Track™ shelving.
  • Track sets include necessary sections of track for assembling track runs up to 21 ft. (640 cm)
  • Coated with Microban™ antimicrobial protection
Order Info
19-138-228 (TTS2.5NA) includes a joining kit to connect the track to other track sets.
Specifica
Product Type Track Set
Da utilizzare con (apparecchiatura) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ top track shelving
Include Necessary sections of track for assembling track runs of 1830 to 6405mm
Lunghezza (sistema imperiale) 168 in.
Lunghezza (sistema metrico) 426.7 cm