missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MFN1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marca:  Invitrogen™ PA598602

Codice prodotto. 16565260

  • 464.00€ / 100µL

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

MFN1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Mitofusin 1 (MFN1) and the related protein MFN2 are mitochondrial membrane GTPase proteins that play a central role in mitochondrial metabolism and may be associated with obesity and/or apoptosis processes. MFN1 and MFN2 form homotypic and heterotypic complexes and coordinately regulate mitochondrial fusion and are essential for embryonic development. When ectopically expressed, MFN1 inhibits the apoptosis-associated amino-terminal conformation change in the apoptotic protein Bax but not its mitochondrial translocation, indicating that MFN1 is involved in the regulating the activation of Bax on the outer mitochondrial membrane.
TRUSTED_SUSTAINABILITY
Specifica

Specifica

MFN1
Polyclonal
Unconjugated
MFN1
2310002F04Rik; 6330416C07Rik; D3Ertd265e; DKFZp762F247; EC 3.6.5; FLJ20693; Fzo homolog; Fzo1b; hfzo1; hfzo2; HR2; Mfn1; MFN2; MGC41806; mitochondrial mitofusin 1; mitochondrial transmembrane GTPase Fzo-1; mitochondrial transmembrane GTPase FZO1B; mitochondrial transmembrane GTPase FZO-2; mitofusin 1; mitofusin 2; mitofusin-1; mKIAA4032; putative transmembrane GTPase; transmembrane GTPase MFN1
Rabbit
Antigen affinity chromatography
RUO
55669, 67414
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q811U4, Q8IWA4
MFN1
Recombinant Human Mitofusin-1 protein (622-741AA).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.