Learn More
MFSD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 21037-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| MFSD7 | |
| Polyclonal | |
| Unconjugated | |
| MFSD7 | |
| LP2561, MFSD7, MYL5, UNQ385/PRO717 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 243197, 305625, 84179 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UXD7, Q8CE47 | |
| MFSD7 | |
| MFSD7 Fusion Protein Ag15361 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |