Learn More
MHC Class II (I-A/I-E) Rat anti-Mouse, FITC, Clone: M5/114.15.2, Proteintech
Rat Monoclonal Antibody
Marca: Proteintech FITC-65122-100UG
Descrizione
The MHC Class II (I-A/I-E) antibody from Proteintech is a rat monoclonal antibody generated with activated c57bl/6 mouse spleen cells of mouse MHC Class II (I-A/I-E). This antibody recognizes mouse antigen. The MHC Class II (I-A/I-E) antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifica
| MHC Class II (I-A/I-E) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| M5/114.15.2 | |
| FITC | |
| 100 μg | |
| 14960 | |
| IgG2b |