missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MHC II Monomorphic Monoclonal Antibody (21-1A6), FITC

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA528490
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MHC II Monomorphic Monoclonal Antibody for Flow
MHC II Monomorphic is part of major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifica
| MHC II Monomorphic | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.09% sodium azide | |
| 0 | |
| BLB1, BLB2, BLB3 | |
| Chicken bursa cells. | |
| 100 μg | |
| Primary | |
| Chicken | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 21-1A6 | |
| FITC | |
| BLB2 | |
| beta 1 domain; B-L; B-L beta chain; B-L beta minor; BLA-DRB3; B-LB; BLB major; BLB1; BLB2; B-LB2; B-LB21; B-LB21 major; BLB3; Bl-Beta II protein; B-LBII; BoLA-DQA; BoLA-DQA1; BOLA-DQA2; BOLA-DQA2 protein; BOLA-DQA3; BOLA-DQA5; BOLA-DQB; BOLA-DRA; BoLA-DR-alpha; BoLA-DRB; BOLA-DRB3; BoLA-DRB3 protein; BoLA-DRB3.2; Bota-DRB01; Bota-DRB02; Bota-DRB04; Bota-DRB07; Bota-DRB21; Bota-DRB22; bovine leukocyte antigen; class II histocompatibility antigen, B-L beta chain; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DQA; DLA-DQA1; DLA-DQB; DLA-DQB1; DLA-DR beta; DLA-DRB1; DLA-DRBB1; dog leucocyte antigen DQA; DQA; dqa 7; dqa1; DQALPHA; DQB; dqb1; DR beta-chain antigen binding domain; DRA; DRB; DRB1; DRB3; DRBB1; DR-beta chain; DR-beta chain MHC class II; ELA-DQA; ela-DRA; Eqca-DQA1; Eqca-DQA2; Eqca-DQA3; Eqca-DQB1; Eqca-DQB2; Eqca-DRA; Eqca-DRB1; Eqca-DRB2; Eqca-DRB3; GSP-BLB1; GSP-BLB2; histocompatibility complex, class II, DR beta 3; HLA-DRB1; integral membrane glycoprotein; integral membrane protein; LA-DRB; leukocyte antigen DRB3; major histocompatability complex class II; major histocompatibility class II antigen B-L beta; major histocompatibility complex; major histocompatibility complex class II B; major histocompatibility complex class II beta chain; Major histocompatibility complex class II beta chain BLB1; Major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB2, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB3, (similar to HLA class II, D beta chain); major histocompatibility complex class II DR-beta chain; major histocompatibility complex, class II, DQ alpha; major histocompatibility complex, class II, DQ alpha 1; major histocompatibility complex, class II, DQ alpha 1 precursor; major histocompatibility complex, class II, DQ alpha 2; major histocompatibility complex, class II, DQ alpha 5; major histocompatibility complex, class II, DQ alpha, type 1; major histocompatibility complex, class II, DQ alpha, type 3; major histocompatibility complex, class II, DQ beta; major histocompatibility complex, class II, DQ beta 1; major histocompatibility complex, class II, DR alpha; major histocompatibility complex, class II, DR alpha precursor; major histocompatibility complex, class II, DR beta 3; major histocompatibility complex, class II, DR beta 4 precursor; major histocompatibility complex, class II, DRB3; MHC class II antigen; MHC class II antigen alpha chain; MHC class II antigen beta chain; MHC class II antigen B-F minor heavy chain; MHC class II antigen BoLA-DRB3; MHC class II antigen precursor; MHC Class II beta 1 and 2 domains; MHC class II beta 1 domain; MHC class II beta chain; MHC class II beta chain 1; MHC class II beta chain 2; MHC class II B-L beta chain; MHC class II DLA DQ beta chain; MHC class II DLA DQA alpha chain; MHC class II DLA DQalpha chain; MHC class II DLA DRB1 beta chain; MHC class II DLA-DRB; MHC class II DLA-DR-beta-1; MHC class II DQA; MHC class II DQA precursor; MHC class II DQA1 precursor; MHC class II DQA2 (BoLA-DQA2); MHC class II DQA2 precursor; MHC class II DQ-alpha; MHC class II DQ-alpha chain; MHC class II DQB precursor; MHC class II DQ-beta chain; MHC class II DR alpha chain; MHC class II DR beta 1; MHC class II DR beta chain; MHC class II DR beta-chain; MHC class II DR-alpha; MHC class II DRB3; MHC class II DRB3 precursor; MHC class II DR-beta chain; MHC class II HLA-DQ-alpha; MHC class II leukocyte antigen BoLA-DRB3; MHC class IIA antigen; MHC DLA-DR-beta-1 precursor; MHC(BoLA) class II DR-beta chain; MHCII beta chain; S19; similar to MHC class II DQA2 (BoLA-DQA2); uncharacterized protein LOC101747454; Unknown (protein for MGC:127488); Unknown (protein for MGC:137100) | |
| Mouse | |
| Protein G | |
| RUO | |
| 101747454, 425256, 693256 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto