Learn More
Texwipe™ Micro Absorbond™ ESD Swab
Sfoglia Tutto Texwipe Prodotti
Click to view available options
Quantità:
500
Dimensione della confezione:
pezzi 500
Descrizione
- Hydroentangled continuous-filament polyester head is extremely low in nonvolatile residue
- A compact, electrically dissipative handle and flexible internal head paddle provide support and control
- The softness of the Absorbond™ head also makes it a good choice for sensitive surfaces including coated optics
- The handle is made from a Stat-Rite™ polymer alloy using a patented Inherently Dissipative Polymer (IDP) to provide clean, permanent ESD protection without contributing particulate contaminates to the cleanroom
- Thermally bonded head
- Exceptionally soft and nonabrasive
- Ultralow nonvolatile residue
- Low particle generation
- Good sorbency
- No contaminating adhesives
- ESD control without contaminating carbon or metal loading
- No blooming ionic antistats
- No contaminating coatings
- Humidity-independent ESD control for consistent performance
- Safely dissipates charges in under two seconds when used with a solvent such as IPA Applications
- Magneto-resistive (MR) head manufacturing and assembly
- Cleaning with solvents such as IPA
- Applying lubricants and other liquids
- Removing excess materials
- Wiping sensitive surfaces including coated optics
Specifica
Specifica
| Quantità | 500 |
| Materiale maniglia | Stat-rite ESD, Acrylic Polymer |
| Tip Material | Hydroentangled Continuous-filament Polyester |