Learn More
Microbiologics™ Staphylococcus aureus subsp. aureus ATCC™ 6538™†
Codice prodotto. 14813271
Encompass_Preferred
Click to view available options
Product Type:
KWIK-STIK
Quantità:
6/Pk.
Dimensione della confezione:
pezzi 6
Codice prodotto. 14813271
Marca: Microbiologics™ 0485K
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Staphylococcus aureus subsp. aureus in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2, 6 or 10
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of microorganism
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 or 10 pellets
  • ATCC™ 6538™†
Specifica
ATCC® 6538
Livello di biosicurezza 2
Certificazione/Conformità CE, online certificate of analysis
Content And Storage 2°C to 8°C
Forma Lyophilized Pellet
Tipo di media Kwik-stik™
Product Type KWIK-STIK
Da utilizzare con (applicazione) Culture purpose and QC application
Quantità 6/Pk.
Specie Staphylococcus aureus
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.