Learn More
Staphylococcus epidermidis ATCC™ 12228™†, Microbiologics™
Codice prodotto. p-4395432
Encompass
Click to view available options
Quantità:
2/Pk.
Product Type:
KWIK-STIK
Dimensione della confezione:
pezzi 2
Codice prodotto. 11986645
Marca: Microbiologics 0371P
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Staphylococcus epidermidis in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2, 6 or 1
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of Staphylococcus epidermidis
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 or 10 pellets
  • ATCC™ 12228™†
Specifica
ATCC® ATCC 12228
Livello di biosicurezza 1
Certificazione/Conformità CE, online certificate of analysis
Content And Storage 2°C to 8°C
Forma Lyophilized Pellet
Tipo di media Kwik-stik™
Product Type KWIK-STIK
Da utilizzare con (applicazione) Culture purpose and QC application
Quantità 2/Pk.
Specie Staphylococcus epidermidis
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.