Learn More
Ansell MICROFLEX™ MEGA TEXTURE 93-283 Blue Gloves
Click to view available options
Size:
Large
3X-Large
Dimensione della confezione:
pezzi 44
pezzi 500
Descrizione
- Mega Texture surface for secure grip when handling slippery objects
- Generous fit works well with thermal under gloves
- Cut-protective design resists tearing
- Compliant with US Food Contact and EU Food Migration Regulations
Specifica
Specifica
| Certificazione/Conformità | ASTM D6319, FDA 21 CFR 177-2600, ISO 9001, ISO 13485 |
| Certifications/Compliance (Europe) | EN 374-1:2016, EN 420:2003 + A1:2009, EN 374-5:2016 |
| Chemotherapeutic Rated | No |
| Accelerator Free | No |
| Applicazioni | Industrial |
| AQL | 1.5 |
| Colore | Blue |
| Monouso | Single-use |
| Finitura esterna | Fully Textured |
| Latex Free | Sì |
| Vedi altri risultati |