Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassette in Quickload™ stacks
Secure specimens during processing with Simport Scientific™ Micromesh™ Biopsy Processing/Embedding Cassette. No need for biopsy pads thanks to the 1676 square 0.36mm openings of the mesh.
Specifica
Materiale Acetal
Quantità 2000
Visualizza altre specifiche
Descrizione
Descrizione
In Quickload™ stacks
  • 45° Anterior writing area
  • 1676 square opening (0.36 mm)
  • No biopsy pads needed
  • 1 large 27 mm compartment
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifica
Specifica
Acetal
2000