Learn More
Micronova™ M-Zone™ LoopMop™ String Mop
GREENER_CHOICE
Codice prodotto. 11986865 Sfoglia Tutto Micronova Prodotti
Click to view available options
Materiale:
Polyester, Polyester (Band)
Dimensione della confezione:
pezzi 24
Codice prodotto. 11986865
Marca: Micronova™ MZLM512
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Requires
MH-1 or MHC-1, or QDMH-1 frame with SSU series handle
A knitted polyester, tubular looped string mop that is designed to effectively lay down disinfectant for the required contact time in pharmaceutical, aseptic processing and medical device applications.
The Micronova M-Zone™ LoopMop™ is a functional, 100% knitted polyester string mop that features a looped design with no exposed raw edges and tubular strands that draw up moisture efficiently and ensure adequate disinfectant dispersal. The durable construction of the mop allows it to be laundered, autoclaved if necessary, and reused.
  • 100% knitted polyester, tubular looped string mop
  • Tubular strands effectively lay down disinfectant for the required contact time
  • Looped design with no exposed raw edges
  • Designed for pharmaceutical production, medical device and cleanroom use
  • For use with the MH-1 Mop Handle or attached to the SSU series handle via the QDMH-1 Mop Adapter
  • Available gamma irradiated for aseptic areas
  • Launderable for reuse
  • Autoclavable
Specifica
Dimensions 38.1 x 17.8 cm (15 x 7 in.)
Product Type String Mop
Colore Bianco
Lunghezza (sistema imperiale) 15 pollici
Sterilità Non-sterile
Lunghezza (sistema metrico) 38.1 cm
Autoclavabile Sterilizzabile in autoclave
Larghezza (sistema metrico) 17.8 cm
Larghezza (sistema imperiale) 7 in.
Resistente a Prodotti chimici
Monouso Reusable
Quantità 24/Cs.
Care Instructions Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
Da utilizzare con (applicazione) Floor Cleaning and Sanitization in Cleanrooms and Controlled Environments
Color-coded No
Materiale Polyester, Polyester (Band)