Learn More
Ismatec™ Micropump™ Gear Pump Head 185
Codice prodotto. 11973124
Click to view available options
Portata:
1 to 100 mL/min
5 to 504 mL/min
Dimensione della confezione:
Pezzo
Codice prodotto. 11973124
Marca: Ismatec™ 7400423
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe design provides higher delivery pressures and temperatures, plus increased efficiency.
  • Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery.
  • The magnetic drive and static seals keep fluids securely inside and potential contaminants out.
  • Chemical resistant materials handle a wide range of materials (due to wetted parts).
  • Pump heads are compact and easy to service.
  • The modular design offers flexibility—configure to your specific needs.
Specifica
Portata 1 to 100 mL/min