Learn More
Ismatec™ Micropump™ Gear Pump Head 200
Codice prodotto. p-8010686
Click to view available options
Portata:
100 to 2016mL/min
197 to 3924 mL/min
Dimensione della confezione:
Pezzo
Codice prodotto. 11964888
Marca: Ismatec™ 00133VW
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe design provides higher delivery pressures and temperatures, plus increased efficiency.
  • Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery.
  • The magnetic drive and static seals keep fluids securely inside and potential contaminants out.
  • Chemical resistant materials handle a wide range of materials (due to wetted parts).
  • Pump heads are compact and easy to service.
  • The modular design offers flexibility—configure to your specific needs.
Specifica
Portata 100 to 2016mL/min