Learn More
Eppendorf™ Tube Racks
Sfoglia Tutto Eppendorf Prodotti
Click to view available options
Posizioni:
1
12
12 (6 and 6)
36
48
Da utilizzare con (apparecchiatura):
0.5 mL Tubes
1 Conical Tube 25 mL or 50 mL
1.5 and 2.0 mL Tubes
5 mL and 15 mL Tubes
5 or 15 mL, 25 or 50 mL Tubes
Cryogenic Tubes
Dimensione della confezione:
pezzi 2
pezzi 4
Descrizione
- Stackable within one rack format and also between all rack variants; saves laboratory space when not in use
- Non-slip silicone feet and a tilt-proof footprint design ensure safe handling
- Wavelike design: Tube rows are arranged in different heights so that the reaction vessels are easily accessible and clearly identifiable
- Made of polypropylene (PP) for high chemical and physical resistance
- Optimized space-saving dimensions due to perfect tube position-to-footprint arrangement without compromising the tube handling
- High flexibility: holds tubes of different sizes
- Numbered tube positions support fast and easy orientation and error-free handling
Specifica
Specifica
| Product Type | Test Tube Stand |
| Capacità | PCR Tubes |
| Materiale | Polypropylene |
| Posizioni | 36 |
| Sovrapponibile | Sì |
| Da utilizzare con (apparecchiatura) | Cryogenic Tubes |
| N. per confezione | 2 Pk. |