Learn More
SPEX CertiPrep™ Microvial Set
Codice prodotto. 15380472 Sfoglia Tutto SPEX Certiprep Prodotti
Click to view available options
Capacità (sistema metrico):
0.1 to 0.5 mL
Dimensione della confezione:
1 set
Codice prodotto. 15380472
Marca: SPEX CertiPrep™ 6757
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Ideal for grinding very small quantities of samples such as plant and animal tissue, seeds, textiles, pharmaceuticals, hair, and bones.
These microvial sets for use with the SPEX SamplePrep freezer mills are ideal for grinding very small quantities of samples such as plant and animal tissue, seeds, textiles, pharmaceuticals, hair, and bones. It is comprised of three microvials contained in a vial holder to allow simultaneous sample processing of three samples up to 0.5g and from 0.1 to 0.5mL.
  • Grinds three samples simultaneously in the 6870 freezer mill, when used in conjunction with 6806 multi-vial adapter
  • Sample grinding capacity 0.1 - 0.5 grams
  • Withstands liquid nitrogen temperatures
  • Closed grinding vial prevents sample cross contamination, retains volatiles and controls hazardous or critical samples
  • Unique vial holder allows liquid nitrogen to flow freely around each microvial
  • 6753C microvial sample extraction tool aids in the removal of ground samples from centre cylinders
Specifica
Capacità (sistema metrico) 0.1 to 0.5 mL
Descrizione Grinds three samples at a time, when used in conjunction with 6806 multi-vial adapter
Quantità confezione 3 Microvials
Peso (sistema metrico) 0.1 to 0.5 g