Learn More
Merck Millicell™ Receiver Trays
Sfoglia Tutto Merck Prodotti
Click to view available options
Conteggio pozzetti:
1
24
Unit Size:
Pezzo
pezzi 5
Description
Designed to support cell attachment, growth, and differentiation for a variety of applications
- 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
- Single-well feeder tray utilizes baffles to reduce media leakage and contamination
Specifications
Specifications
| Chiusura inclusa | With Lid |
| Colore | Clear |
| Materiale | Polistirene |
| Forma | Rectangular |
| Sterilità | Sterile |
| Tipo | Receiver Tray |
| Conteggio pozzetti | 24 |
| Include | 24-Well Receiver Tray, Lid |
| Membrana di supporto | PCF |