Learn More
BOLA Miniature 3-Way Stopcocks
Click to view available options
Adatto al tubo ID:
2 connections
3 connections
T shaped
Diametro (sistema metrico):
0.8 x 1.6 mm
1.6 x 3.2 mm
Dimensione della confezione:
Pezzo
Descrizione
- 3-way stopcock with L-shaped or T-shaped bore and three connections.
- Available either with three female threads UNF ¼ in. 28 G or with two female threads UNF ¼ in. 28 G and one male thread UNF ¼ in. 28 G.
- Total height including stopcock plug made of FEP: 20 mm.
Specifica
Specifica
| Diametro (sistema metrico) | 0.8 x 1.6 mm |
| Autoclavabile | Autoclavable |
| Adatto al tubo ID | 3 connections |
| Lunghezza (sistema metrico) | 32 mm |
| Certificazione/Conformità | FDA |
| Connectors | 3-way |