Learn More
Kinematica™ Mixing Attachment for Microtron™ MB 550 Laboratory Mixer
Click to view available options
Capacità (sistema metrico):
125 mL
250 ml
500 mL
1000 mL
Dimensione della confezione:
Pezzo
Descrizione
- For use with the MB 550 Mixers
- The two stainless steel ball bearings are sealed twice each
- Noiseless and sturdy coupling with dampers
- Blades, sealing and dampers can easily be exchanged
- Four high-grade blades designed for optimal mixing
- Generally suited for intensive and fast processing
Specifica
Specifica
| Product Type | Mixing Head Attachment |
| Capacità (sistema metrico) | 500 mL |
| Materiale della lama | Stainless Steel |
| Materiale | Borosilicate Glass |
| Da utilizzare con (apparecchiatura) | MB 550 Laboratory Mixer |
| Include | Gasket and blade head |
| Quantità confezione | 1 Ea. |
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering