Learn More
Fisherbrand™ TV400YK-MODSYS Complete Modular System
Click to view available options
Tipo:
Complete Modular System
Dimensione della confezione:
Pezzo
Descrizione
- Complete systems that offer the maximum in space saving and economy.
- Complete 2D analysis and blotting
- Comprises cooled vertical tank complete with casting base and accessories plus the 1D module, plus the electroblotting module and accessories
Specifica
Specifica
| Tipo | Complete Modular System |
| Include | With GRM, cooled gel tank and quick-fit tubing, lid, 2 (20.5 x 20cm; W x H) plain glass plates, 4 x 1mm spacers, 2 x spacer aligners, 2 (20.5 x 20cm) notched glass plates, 1 x dummy plate, 2 x 1mm thick 24-sample combs, casting base and 2 x silicone seals; CAPGRM, 10 x 1mm glass capillaries, 4 x 1.5mm thick spacers, 4 x 1.5mm thick 1-D preparatory combs, 10 x blanking ports and 1 x gel extraction platform; plus EBGRM, 2 x electroblotting cassettes and 4 x fibre pads |
| Dimensioni (L x P x A) | D x W x H: 15 x 38 x 27.5 cm |
| Requisiti elettrici | 200/300 V (10 to 15 V/cm) 30/40 mA |