missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moesin Antibody (MSN/492), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-47916PECY55
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Moesin Monoclonal specifically detects Moesin in Human, Rat (Negative) samples. It is validated for Flow Cytometry, Knockout Validated.
Specifica
| Moesin | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| MSN | |
| Recombinant full-length human Moesin protein (Uniprot: P26038 ) | |
| 0.1 mL | |
| Primary | |
| Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry, Immunoassay | |
| MSN/492 | |
| Flow Cytometry, Knockout Validated | |
| Membrane-organizing extension spike protein, moesin | |
| Mouse | |
| Protein A or G purified | |
| Cytoskeleton Markers, Stem Cell Markers | |
| 4478.0 | |
| Human, Rat (Negative) | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto