Learn More
Honeywell™ Molded Cup Facemask 5211 XL-FFP2 NR D with Valve and Full-Face Seal
Click to view available options
:
Formato: X-Large
Descrizione
- Rigid shell: shape maintained even in moistened and heavy loading environments.
- Ultra-soft hypoallergenic Willtech™ flange seal with absorption properties and a tight-sealing film for an exceptional comfort and a perfect fit.
- Preformed and colored nosebridge to save time to perfectly adjust the mask and to instantly identify its protection level.
- Large internal volume to easily communicate and breathe.
- Shielded, high performance exhalation valve with an exclusive design (butterfly opening-closing system) for improved protection, comfort, and decreased fatigue.
- Adjustable braids to easily and efficiently fit mask.
- Lightweight respirator (26 g) to reduce fatigue.
- No latex, no silicone, no PVC.
- Dolomite tested.
Specifica
Specifica
| Certificazione/Conformità | EC Category PPE: 3, QA: ISO 9001/2000, EC Attestation, EN 149:2001+A1:2009 |
| Monouso | Single-use |
| Product Type | Rigid Shell |
| Size | X-Large |
| With Exhalation Valve | Sì |
| Da utilizzare con (applicazione) | Agriculture, automotive, construction, food, iron/steel, mining, textile, wood industries |
| Include | Valve, adjustable braids, full-face seal |
| Senza lattice | Yes |
| Quantità | 5/Box, 50/Pk. |