Learn More
Micronova™ Mop LoPro controlled environment NovaPoly 406mm MegaSwat
Sfoglia Tutto Micronova Prodotti
Descrizione
- NovaPoly cover for use with MegaSwat™ mop
- Ideal for cleaning around heavy process equipment or calibrated apparatus
- 100% smooth knit polyester
- Compatible with strong disinfectants
- Autoclavable
Specifica
Specifica
| Sterilità | Non-sterile |