Learn More
MOPS-EDTA-sodium acetate buffer (MESA), Thermo Scientific™
Codice prodotto. 15412999
Cambia vista
Click to view available options
Quantità:
500 mL
1 L
Dimensione della confezione:
1L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15412999 500 mL 500mL
15422999 1 L 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15412999 Fornitore Thermo Scientific Alfa Aesar N. del fornitore J60184.AP
Richiesta di formato massivo o personalizzato
Please to purchase this item. Need a web account? Register with us today!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico MOPS-EDTA-sodium acetate buffer (MESA)
Forma fisica Liquid
Quantità 500 mL
Numero MDL MFCD00131625