missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-CRYAB, Clone: S4, Abnova™
Marca: Abnova H00001410-M01A.200uL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| CRYAB | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC007008.1 | |
| CRYAB | |
| CRYAB (AAH07008.1,1 a.a. ∽ 175 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1410 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| Western Blot | |
| S4 | |
| Mouse monoclonal antibody raised against a full-length recombinant CRYAB. | |
| CRYAB | |
| CRYA2/CTPP2/HSPB5 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction