Learn More
Mouse anti-Human IgG F(ab) Secondary Antibody (4A11), Janelia Fluor 646, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB500-467JF646
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG F(ab) Monoclonal antibody specifically detects IgG F(ab) in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifica
| IgG F(ab) | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 4A11 | |
| 50mM Sodium Borate | |
| Purified human IgG. | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG2b |