Learn More
Mouse anti-Human IgM Heavy Chain Secondary Antibody (47.2.E9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-68319UV
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgM Heavy Chain Monoclonal antibody specifically detects IgM Heavy Chain in Human samples. It is validated for ELISA
Specifica
| IgM Heavy Chain | |
| Monoclonal | |
| DyLight 350 | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Purified human IgM | |
| 0.2 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| 47.2.E9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Secondary | |
| Human | |
| Purified |